Anti-PLD4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-13770
103286-116EA 734.58 USD
103286-116
Anti-PLD4 Rabbit Polyclonal Antibody
Antibodies
The PLD4 / Phospholipase D4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PLD4 / Phospholipase D4. This antibody reacts with human. The PLD4 / Phospholipase D4 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PLD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR