Anti-PLCXD3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-13769
103286-114EA 734.58 USD
103286-114
Anti-PLCXD3 Rabbit Polyclonal Antibody
Antibodies
The PLCXD3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PLCXD3. This antibody reacts with human. The PLCXD3 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PLCXD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR