Anti-PLCG2 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00005336-B01P
103307-586EA 399 USD
103307-586
Anti-PLCG2 Mouse Polyclonal Antibody
Antibodies
The PLC-gamma 2 Antibody from Novus Biologicals is a mouse polyclonal antibody to PLC-gamma 2. This antibody reacts with human. The PLC-gamma 2 Antibody has been validated for the following applications: Western Blot, Proximity Ligation Assay.

Type: Primary
Antigen: PLCG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR