Anti-PLCG1 Mouse Monoclonal Antibody [clone: 2A2]

Supplier: Novus Biologicals
H00005335-M01
103307-584EA 798.46 USD
103307-584
Anti-PLCG1 Mouse Monoclonal Antibody [clone: 2A2]
Antibodies
The PLC-gamma 1 Antibody (2A2) from Novus Biologicals is a mouse monoclonal antibody to PLC-gamma 1. This antibody reacts with human. The PLC-gamma 1 Antibody (2A2) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Proximity Ligation Assay.

Type: Primary
Antigen: PLCG1
Clonality: Monoclonal
Clone: 2A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR