|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | PLBD2 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
ImmunoChemistry | Yes |
Antigen Synonyms | phospholipase B domain-containing protein 2, lamina ancestor homolog 2, LAMA-like protein 2,76 kDa protein, phospholipase B-like 2 45 kDa form, mannose-6-phosphate protein associated protein p76, phospholipase B-like 2 32 kDa form, P76, phospholipase B domain containing 2, PLB homolog 2, putative phospholipase B-like 2 |
Gene ID | 196463 |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:GDWFSYDGSPRAQIFRRNQSLVQDMDSMVRLMRYNDFLHDPLSLCKACNPQPNGENAISARSDLNPANGSYPFQALRQRSHGGIDVKVTSMSLARILSLLAASGPTWDQVPPFQWSTS |
Purification | Immunogen affinity purified |
Storage Buffer | PBS (pH 7.2) and 40% Glycerol |
Storage Temperature | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |