Anti-PLAP Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-19858
102710-588EA 635.51 USD
102710-588
Anti-PLAP Rabbit Polyclonal Antibody
Antibodies
The PLAP Antibody from Novus Biologicals is a rabbit polyclonal antibody to PLAP. This antibody reacts with human. The PLAP Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: PLAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR