Anti-PLAP Mouse Monoclonal Antibody [clone: 1F7]

Supplier: Novus Biologicals
H00009373-M04
103336-736EA 798.46 USD
103336-736
Anti-PLAP Mouse Monoclonal Antibody [clone: 1F7]
Antibodies
The PLAP Antibody (1F7) from Novus Biologicals is a mouse monoclonal antibody to PLAP. This antibody reacts with human. The PLAP Antibody (1F7) has been validated for the following applications: ELISA.

Type: Primary
Antigen: PLAP
Clonality: Monoclonal
Clone: 1F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Lambda
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR