Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PLAGL1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
The PLAGL1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PLAGL1. This antibody reacts with human. The PLAGL1 Antibody has been validated for the following applications: ELISA.

Type: Primary
Antigen: PLAGL1
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg 28830002-0.1MG 103317-018 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol PLAGL1
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Antigen Synonyms ZAC1, pleiomorphic adenoma gene-like 1, pleiomorphic adenoma gene-like protein 1, ZAC tumor supressor, PLAG-like 1, LOT-1, MGC126275, LOT1ZACLost on transformation 1, MGC126276, zinc finger protein PLAGL1, DKFZp781P1017, Pleiomorphic adenoma-like protein 1, Tumor supressor ZAC
Gene ID 5325
Immunogen This antibody is specific for the Middle Region of the target protein.
Purification Immunogen affinity purified
Storage Buffer 20mM Potassium Phosphate (pH 7.0) and 0.15M NaCl
Storage Temperature Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty