Anti-PLAG1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-15075
102705-352EA 635.51 USD
102705-352
Anti-PLAG1 Rabbit Polyclonal Antibody
Antibodies
The PLAG1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PLAG1. This antibody reacts with human. The PLAG1 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: PLAG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR