Anti-PLAG1 Mouse Monoclonal Antibody [clone: 3B7]

Supplier: Novus Biologicals
H00005324-M02
103307-572EA 399 USD
103307-572
Anti-PLAG1 Mouse Monoclonal Antibody [clone: 3B7]
The PLAG1 Antibody (3B7) from Novus Biologicals is a mouse monoclonal antibody to PLAG1. This antibody reacts with human. The PLAG1 Antibody (3B7) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry.

Type: Primary
Antigen: PLAG1
Clonality: Monoclonal
Clone: 3B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR