Anti-PLA2G7 Mouse Monoclonal Antibody [clone: 5B9]

Supplier: Novus Biologicals
H00007941-M01
103334-532EA 798.46 USD
103334-532
Anti-PLA2G7 Mouse Monoclonal Antibody [clone: 5B9]
Antibodies
The PLA2G7 / PAF-AH / Lp-PLA2 Antibody (5B9) from Novus Biologicals is a mouse monoclonal antibody to PLA2G7 / PAF-AH / Lp-PLA2. This antibody reacts with human. The PLA2G7 / PAF-AH / Lp-PLA2 Antibody (5B9) has been validated for the following applications: ELISA.

Type: Primary
Antigen: PLA2G7
Clonality: Monoclonal
Clone: 5B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR