Anti-PLA2G6 Mouse Polyclonal Antibody

H00008398-B01
103334-870EA 583.08 USD
103334-870
Anti-PLA2G6 Mouse Polyclonal Antibody
The PLA2G6 Antibody from Novus Biologicals is a mouse polyclonal antibody to PLA2G6. This antibody reacts with human. The PLA2G6 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: PLA2G6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR