Anti-PLA2G6 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00008398-D01P
103334-874EA 844.62 USD
103334-874
Anti-PLA2G6 Rabbit Polyclonal Antibody
Antibodies
The PLA2G6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PLA2G6. This antibody reacts with human, mouse. The PLA2G6 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: PLA2G6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR