Anti-PLA2G4E Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-32610
103282-492EA 734.58 USD
103282-492
Anti-PLA2G4E Rabbit Polyclonal Antibody
Antibodies
The PLA2G4E Antibody from Novus Biologicals is a rabbit polyclonal antibody to PLA2G4E. This antibody reacts with human. The PLA2G4E Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PLA2G4E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR