Anti-PLA2G4C Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00008605-B01P
103335-296EA 798.46 USD
103335-296
Anti-PLA2G4C Mouse Polyclonal Antibody
Antibodies
The PLA2G4C Antibody from Novus Biologicals is a mouse polyclonal antibody to PLA2G4C. This antibody reacts with human. The PLA2G4C Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: PLA2G4C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR