Anti-PLA2G4B Mouse Monoclonal Antibody [clone: 1G5]

Supplier: Novus Biologicals
H00008681-M01
103335-426EA 798.46 USD
103335-426
Anti-PLA2G4B Mouse Monoclonal Antibody [clone: 1G5]
Antibodies
The PLA2G4B Antibody (1G5) from Novus Biologicals is a mouse monoclonal antibody to PLA2G4B. This antibody reacts with human. The PLA2G4B Antibody (1G5) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PLA2G4B
Clonality: Monoclonal
Clone: 1G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR