Anti-PKR Mouse Monoclonal Antibody [clone: 1B9]

H00005610-M01
103308-294EA 583.08 USD
103308-294
Anti-PKR Mouse Monoclonal Antibody [clone: 1B9]
The PKR Antibody (1B9) from Novus Biologicals is a mouse monoclonal antibody to PKR. This antibody reacts with human. The PKR Antibody (1B9) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PKR
Clonality: Monoclonal
Clone: 1B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR