Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PKR Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-56399SS
103359-018
Anti-PKR Rabbit Polyclonal Antibody
The PKR Antibody from Novus Biologicals is a rabbit polyclonal antibody to PKR. This antibody reacts with human. The PKR Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: PKR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty