Anti-PKNOX1 Mouse Monoclonal Antibody [clone: 2A12]

Supplier: Novus Biologicals
H00005316-M07
103307-560EA 798.46 USD
103307-560
Anti-PKNOX1 Mouse Monoclonal Antibody [clone: 2A12]
Antibodies
The PKNOX1 Antibody (2A12) from Novus Biologicals is a mouse monoclonal antibody to PKNOX1. This antibody reacts with human. The PKNOX1 Antibody (2A12) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: PKNOX1
Clonality: Monoclonal
Clone: 2A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR