Anti-PKN3 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00029941-B01P
103343-162EA 798.46 USD
103343-162
Anti-PKN3 Mouse Polyclonal Antibody
Antibodies
The PKN3 Antibody from Novus Biologicals is a mouse polyclonal antibody to PKN3. This antibody reacts with human. The PKN3 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PKN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR