Anti-PKN1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-56159
103358-684EA 318.77 USD
103358-684
Anti-PKN1 Rabbit Polyclonal Antibody
Antibodies
The PKN1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PKN1. This antibody reacts with human, mouse, rat, canine. The PKN1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PKN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Canine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR