Anti-PKIG Mouse Monoclonal Antibody [clone: 2D9]

Supplier: Novus Biologicals
H00011142-M10
103340-226EA 798.46 USD
103340-226
Anti-PKIG Mouse Monoclonal Antibody [clone: 2D9]
Antibodies
The PKIG Antibody (2D9) from Novus Biologicals is a mouse monoclonal antibody to PKIG. This antibody reacts with human. The PKIG Antibody (2D9) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PKIG
Clonality: Monoclonal
Clone: 2D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR