Anti-PKHD1L1 Mouse Monoclonal Antibody [clone: 1F5]

Supplier: Novus Biologicals
H00093035-M01
103345-928EA 798.46 USD
103345-928
Anti-PKHD1L1 Mouse Monoclonal Antibody [clone: 1F5]
Antibodies
The PKHD1L1 Antibody (1F5) from Novus Biologicals is a mouse monoclonal antibody to PKHD1L1. This antibody reacts with human. The PKHD1L1 Antibody (1F5) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PKHD1L1
Clonality: Monoclonal
Clone: 1F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR