Anti-PKCI Rabbit Polyclonal Antibody

NBP2-19850
102710-572EA 493.85 USD
102710-572
Anti-PKCI Rabbit Polyclonal Antibody
The PKC iota Antibody from Novus Biologicals is a rabbit polyclonal antibody to PKC iota. This antibody reacts with human. The PKC iota Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: PKCI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR