Anti-PKCB Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00005579-D01P
103308-094EA 429 USD
103308-094
Anti-PKCB Rabbit Polyclonal Antibody
Antibodies
The PKC beta Antibody from Novus Biologicals is a rabbit polyclonal antibody to PKC beta. This antibody reacts with human. The PKC beta Antibody has been validated for the following applications: Western Blot, Proximity Ligation Assay.

Type: Primary
Antigen: PKCB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR