Anti-PKCA Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-87269
103270-540EA 734.58 USD
103270-540
Anti-PKCA Rabbit Polyclonal Antibody
Antibodies
The PKC alpha Antibody from Novus Biologicals is a rabbit polyclonal antibody to PKC alpha. This antibody reacts with human, mouse, rat. The PKC alpha Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PKCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR