Anti-PKA C beta Mouse Monoclonal Antibody [clone: 1F8]

Supplier: Novus Biologicals
H00005567-M02
103308-068EA 399 USD
103308-068
Anti-PKA C beta Mouse Monoclonal Antibody [clone: 1F8]
Antibodies
The PKA C beta Antibody (1F8) from Novus Biologicals is a mouse monoclonal antibody to PKA C beta. This antibody reacts with human. The PKA C beta Antibody (1F8) has been validated for the following applications: ELISA.

Type: Primary
Antigen: PKA C beta
Clonality: Monoclonal
Clone: 1F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR