Anti-PITX2 Mouse Monoclonal Antibody [clone: 2G6]

Supplier: Novus Biologicals
H00005308-M01
103307-514EA 798.46 USD
103307-514
Anti-PITX2 Mouse Monoclonal Antibody [clone: 2G6]
Antibodies
The PITX2 Antibody (2G6) from Novus Biologicals is a mouse monoclonal antibody to PITX2. This antibody reacts with human. The PITX2 Antibody (2G6) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: PITX2
Clonality: Monoclonal
Clone: 2G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR