Anti-PIP5K3 Mouse Monoclonal Antibody [clone: 1D11]

Supplier: Novus Biologicals
H00200576-M02
103347-692EA 798.46 USD
103347-692
Anti-PIP5K3 Mouse Monoclonal Antibody [clone: 1D11]
Antibodies
The PIKFyve / PIP5K3 Antibody (1D11) from Novus Biologicals is a mouse monoclonal antibody to PIKFyve / PIP5K3. This antibody reacts with human. The PIKFyve / PIP5K3 Antibody (1D11) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PIP5K3
Clonality: Monoclonal
Clone: 1D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR