Anti-PIP5K2 gamma Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-19836
102710-544EA 635.51 USD
102710-544
Anti-PIP5K2 gamma Rabbit Polyclonal Antibody
Antibodies
The PIP5K2 gamma Antibody from Novus Biologicals is a rabbit polyclonal antibody to PIP5K2 gamma. This antibody reacts with human. The PIP5K2 gamma Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: PIP5K2 gamma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR