Anti-PIP5K1C Mouse Monoclonal Antibody [clone: 2E9]

Supplier: Novus Biologicals
H00023396-M02
103341-276EA 798.46 USD
103341-276
Anti-PIP5K1C Mouse Monoclonal Antibody [clone: 2E9]
Antibodies
The PIP5K1C Antibody (2E9) from Novus Biologicals is a mouse monoclonal antibody to PIP5K1C. This antibody reacts with human. The PIP5K1C Antibody (2E9) has been validated for the following applications: Western Blot, ELISA, Immunoprecipitation.

Type: Primary
Antigen: PIP5K1C
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR