Anti-PIP5K1C Mouse Monoclonal Antibody [clone: 7D11]

Supplier: Novus Biologicals
H00023396-M10
103341-278EA 798.46 USD
103341-278
Anti-PIP5K1C Mouse Monoclonal Antibody [clone: 7D11]
Antibodies
The PIP5K1C Antibody (7D11) from Novus Biologicals is a mouse monoclonal antibody to PIP5K1C. This antibody reacts with human. The PIP5K1C Antibody (7D11) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PIP5K1C
Clonality: Monoclonal
Clone: 7D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR