Anti-PIP5K1C Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00023396-D01P
103341-274EA 844.62 USD
103341-274
Anti-PIP5K1C Rabbit Polyclonal Antibody
Antibodies
The PIP5K1C Antibody from Novus Biologicals is a rabbit polyclonal antibody to PIP5K1C. This antibody reacts with human. The PIP5K1C Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PIP5K1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR