Anti-PIP Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-83386
103259-150EA 777.05 USD
103259-150
Anti-PIP Rabbit Polyclonal Antibody
Antibodies
The PIP Antibody from Novus Biologicals is a rabbit polyclonal antibody to PIP. This antibody reacts with human. The PIP Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR