Anti-PINK1 Rabbit Polyclonal Antibody (Alexa Fluor® 405)

Supplier: Novus Biologicals
BC100-494AF405
103318-634EA 700.31 USD
103318-634
Anti-PINK1 Rabbit Polyclonal Antibody (Alexa Fluor® 405)
Antibodies
The PINK1 Antibody [Alexa Fluor« 405] from Novus Biologicals is a rabbit polyclonal antibody to PINK1. This antibody reacts with human, mouse, rat, drosophila. The PINK1 Antibody [Alexa Fluor« 405] has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PINK1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 405
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Drosophila
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR