Anti-PIM2 Goat Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-26115
103408-988EA 564.74 USD
103408-988
Anti-PIM2 Goat Polyclonal Antibody
Antibodies
The PIM2 Antibody from Novus Biologicals is a goat polyclonal antibody to PIM2. This antibody reacts with human. The PIM2 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.

Type: Primary
Antigen: PIM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR