Anti-PIK3R3 Rabbit Polyclonal Antibody

NBP1-87366
103271-450EA 607.69 USD
103271-450
Anti-PIK3R3 Rabbit Polyclonal Antibody
The PIK3R3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PIK3R3. This antibody reacts with human. The PIK3R3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PIK3R3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR