Anti-PIK3CA Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-19804
102710-480EA 635.51 USD
102710-480
Anti-PIK3CA Rabbit Polyclonal Antibody
Antibodies
The PIK3CA Antibody from Novus Biologicals is a rabbit polyclonal antibody to PIK3CA. This antibody reacts with human, mouse. The PIK3CA Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PIK3CA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR