Anti-pIgR Mouse Monoclonal Antibody [clone: 2G5]

Supplier: Novus Biologicals
H00005284-M01
103307-426EA 767.69 USD
103307-426
Anti-pIgR Mouse Monoclonal Antibody [clone: 2G5]
Antibodies
The pIgR Antibody (2G5) from Novus Biologicals is a mouse monoclonal antibody to pIgR. This antibody reacts with human. The pIgR Antibody (2G5) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: pIgR
Clonality: Monoclonal
Clone: 2G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR