Anti-PIGO Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-19828
102710-528EA 635.51 USD
102710-528
Anti-PIGO Rabbit Polyclonal Antibody
Antibodies
The PIGO Antibody from Novus Biologicals is a rabbit polyclonal antibody to PIGO. This antibody reacts with human. The PIGO Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PIGO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR