Anti-PIGL Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-86471
103270-250EA 734.58 USD
103270-250
Anti-PIGL Rabbit Polyclonal Antibody
Antibodies
The PIGL Antibody from Novus Biologicals is a rabbit polyclonal antibody to PIGL. This antibody reacts with human. The PIGL Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PIGL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR