Anti-PIGC Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-86283
103272-722EA 734.58 USD
103272-722
Anti-PIGC Rabbit Polyclonal Antibody
Antibodies
The PIGC Antibody from Novus Biologicals is a rabbit polyclonal antibody to PIGC. This antibody reacts with human. The PIGC Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PIGC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR