Anti-PIGC Mouse Monoclonal Antibody [clone: 1G2]

Supplier: Novus Biologicals
H00005279-M01
103307-420EA 798.46 USD
103307-420
Anti-PIGC Mouse Monoclonal Antibody [clone: 1G2]
Antibodies
The PIGC Antibody (1G2) from Novus Biologicals is a mouse monoclonal antibody to PIGC. This antibody reacts with human. The PIGC Antibody (1G2) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PIGC
Clonality: Monoclonal
Clone: 1G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR