|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | PIGA |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
Antigen Synonyms | GPI3, GlcNAc-PI synthesis protein, EC 2.4.1.198, phosphatidylinositol glycan, phosphatidylinositol N-acetylglucosaminyltransferase subunit A, GPI anchor biosynthesis, class A protein, class A, Phosphatidylinositol-glycan biosynthesis class A protein, phosphatidylinositol-glycan biosynthesis, phosphatidylinositol glycan anchor biosynthesis, class A (paroxysmal nocturnal hemoglobinuria), class A GlcNAc-inositol phospholipid assembly protein, PIG-A |
Gene ID | 5277 |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: FADVSSVLTNKLLTVSLCDTNHIICVSYTSKENTVLRAALNPEIVSVIPN AVDPTDFTPDPFRRHDSITIVVVSRLVYRKGIDLLSGIIPELCQKYPDLN FIIGGEGPKRII |
Purification | Immunogen affinity purified |
Storage Buffer | PBS (pH 7.2) and 40% Glycerol |
Storage Temperature | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |