|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | PIG3 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
Antigen Synonyms | p53-induced gene 3 protein, Tumor protein p53-inducible protein 3, PIG3quinone oxidoreductase homolog, tumor protein p53 inducible protein 3, quinone oxidoreductase PIG3 |
Gene ID | 9540 |
Concentration | 1 mg/ml |
Immunogen | Recombinant protein encompassing a sequence within the center region of human PIG3. The exact sequence is proprietary. |
Purification | Immunogen affinity purified |
Storage Buffer | 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol |
Storage Temperature | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |