|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | PIG3 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ELISA | Yes |
Antigen Synonyms | PIG3quinone oxidoreductase homolog, quinone oxidoreductase PIG3, Tumor protein p53-inducible protein 3, p53-induced gene 3 protein, tumor protein p53 inducible protein 3 |
Gene ID | 9540 |
Immunogen | This antibody is specific for the N Terminus Region of the target protein. |
Purification | Immunogen affinity purified |
Storage Buffer | 20mM Potassium Phosphate (pH 7.0) and 0.15M NaCl |
Storage Temperature | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |