Anti-PIG3 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00009540-B02P
103337-198EA 399 USD
103337-198
Anti-PIG3 Mouse Polyclonal Antibody
Antibodies
The PIG3 Antibody from Novus Biologicals is a mouse polyclonal antibody to PIG3. This antibody reacts with human. The PIG3 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PIG3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR