Anti-PIEZO2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-13759
103286-100EA 449 USD
103286-100
Anti-PIEZO2 Rabbit Polyclonal Antibody
The PIEZO2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PIEZO2. This antibody reacts with human. The PIEZO2 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PIEZO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR