Anti-Piccolo Mouse Monoclonal Antibody [clone: 6H9-B6]

Supplier: Novus Biologicals
NBP1-49453SS
102179-038EA 309.97 USD
102179-038
Anti-Piccolo Mouse Monoclonal Antibody [clone: 6H9-B6]
Antibodies
The Piccolo Antibody (6H9-B6) from Novus Biologicals is a mouse monoclonal antibody to Piccolo. This antibody reacts with human, mouse, rat. The Piccolo Antibody (6H9-B6) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: Piccolo
Clonality: Monoclonal
Clone: 6H9-B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR