Anti-Piccolo Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00027445-B01P
103342-848EA 399 USD
103342-848
Anti-Piccolo Mouse Polyclonal Antibody
Antibodies
The Piccolo Antibody from Novus Biologicals is a mouse polyclonal antibody to Piccolo. This antibody reacts with human. The Piccolo Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Piccolo
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR